Learn More
MEDICOM Herkul™ Grip Extra Strong Nitrile Gloves
Artikelnummer. 31781377 Alle ansehen Medicom Produkte
Ansicht ändern
Click to view available options
Größe:
Medium
Large
X-Large
2X-Large
Packungsgröße:
1000 Stück
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Größe unitSize
31781377 X-Large 1000 Stück
31781317 2X-Large 1000 Stück
31781417 Medium 1000 Stück
31781326 Large 1000 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 31781377 Lieferant MEDICOM Lieferanten-Nr. HORNG8N10XL
Verfügbar ab: 21-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
The orange Herkul™ Grip Extra Strong nitrile glove combines enhanced visibility and exceptional 3D grip for intensive automotive and industrial work.
  • Unique 3D textured surface
  • Good grip on oily surface
  • Maximum dexterity
  • Very thick glove for greater durability
  • High level of protection against chemicals (JKPT)
  • Quick donning
  • Fitted cuff for better comfort
Spezifikation
Accelerator Free No
Anwendungen Industrial
Farbe Orange
Einwegprodukt Single-use
Oberflächenbehandlung außen Fully Textured
Finger Thickness (Metric) 0.26 mm
Latex Free Yes
Material Nitrile
Palm Thickness (Metric) 0.23 mm
Gepudert Powder Free
Produkttyp Nitrile Gloves
Größe X-Large
Sterilität Non-sterile
Dicke (metrisch) 0.26 mm