missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HES1 Monoclonal Antibody (960216)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561296
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.
Spezifikation
| HES1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q14469 | |
| HES1 | |
| E. coli-derived recombinant human HES-1, Asn90-Pro277. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 960216 | |
| Unconjugated | |
| HES1 | |
| BHLHB39; class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; hairy and enhancer of split 1 (Drosophila); hairy and enhancer of split 1,; hairy homolog; Hairy-like; Hairy-like protein; hes family bHLH transcription factor 1; HES1; Hes-1; hHL; HL; HRY; RHL; transcription factor HES-1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3280 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur