missing translation for 'onlineSavingsMsg'
Learn More

Hexadecanoic-11,11,12,12-d4 Acid, CDN

Product Code. 30314935
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
0.05 g
0.1 g
missing translation for 'unitSize'
0.05 Gramm
0.10 Gramm
Product Code. Menge unitSize
30314935 0.1 g 0.10 Gramm
30311712 0.05 g 0.05 Gramm
2 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30314935

missing translation for 'mfr': CDN CDND18890.1G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Hexadecanoic-11,11,12,12-D4 Acid
CAS 75736-57-1
Summenformel C16 D4 H28 O2
Menge 0.1 g
Synonym Hexadecanoic-11,11,12,12-d4 acid (9 CI), Hexadecanoic-11,11,12,12-D4 Acid, Palmitic acid-11,11,12,12-d4, [11,11,12,12-2H4]Hexadecanoic acid, [11,11,12,12-2H4]Palmitic acid, Hexadecanoic Acid-11,11,12,12-d4
InChI-Formel InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i5D2,6D2
SMILES [2 H]C([2 H])(CCCC)C([2 H])([2 H])CCCCCCCCCC(=O)O
IUPAC-Name 11,11,12,12-tetradeuteriohexadecanoic acid
Molekulargewicht (g/mol) 260.4487
Formelmasse 260.2653 g/mol
Reinheit (%) 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.