missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
Specifications
| Antigen | HIF-1 alpha |
| Anwendungen | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay |
| Klassifikation | Monoclonal |
| Klon | 2443B |
| Konjugat | DyLight 405 |
| Verdünnung | Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated |
| Zusammensetzung | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gen-Alias | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp |
| Gensymbole | HIF1A |
| Wirtsspezies | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?