missing translation for 'onlineSavingsMsg'
Learn More

His Tag Antibody (6HIS/6402R), Allophycocyanin, Novus Biologicals™

Artikelnummer. 18602027 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18602027 100 μg 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18602027 Leverandør Novus Biologicals Leverandørnr. NBP308360APC

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen His Tag
Anwendungen Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Klassifikation Monoclonal
Klon 6HIS/6402R
Konjugat APC
Zusammensetzung PBS
Gen-Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Wirtsspezies Rabbit
Immunogen Hexa-histidine tagged human CD70 recombinant protein
Reinigungsverfahren Protein A or G purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Cellular Markers, Epitope Tags
Primär oder sekundär Primary
Zielspezies Tag
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.