missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AG Antibody (CL5041) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44252
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HIST1H2AG Monoclonal antibody specifically detects HIST1H2AG in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| HIST1H2AG | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| H2A histone family, member P, H2A.1, H2A.1b, H2A/p, H2AFC, H2AFD, H2AFI, H2AFN, H2AFP, HIST1H2AI, HIST1H2AK, HIST1H2AL, HIST1H2AM, histone 1, H2ag, histone cluster 1, H2ag, histone H2A type 1, Histone H2A/p, pH2A/f | |
| This antibody was generated using a synthetic peptide of P0C0S8, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL5041 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8969 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur