Learn More
Histone H3 Antibody (Immunomedics patent anti-Histone H3), FITC, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spezifikation
Spezifikation
| Antigen | Histone H3 |
| Anwendungen | Flow Cytometry, ELISA, Functional Assay |
| Klassifikation | Monoclonal |
| Klon | Immunomedics patent anti-Histone H3 |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Wirtsspezies | Human |
| Immunogen | Histone H3 |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |