Learn More
HIV-1 Gag p24 Antibody (28), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB100-65515
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).
Spezifikation
| HIV-1 Gag p24 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:10-1:50, ELISA 1:20-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:50, Immunohistochemistry-Paraffin (Negative) | |
| gag | |
| HIV-1 Gag p24 | |
| 0.05 mg | |
| Primary | |
| Specific for the HIV-1 Gag p24, this forms the conical core of the virus that encapsulates the genomic RNA-nucleocapsid complex. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 28 | |
| Unconjugated | |
| Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 155030 | |
| Virus | |
| Purified |