Learn More
HK3 Mouse, anti-Human, Unconjugated, Clone: 3H3C7, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67803-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The HK3 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human HK3. This antibody recognizes human, mouse, rat, pig antigen. The HK3 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Spezifikation
| HK3 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 3101 | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 3H3C7 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |