missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-A2 Monoclonal Antibody (BB7.2), Super Bright™ 600, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
171.25 € - 391.00 €
Spezifikation
| Antigen | HLA-A2 |
|---|---|
| Klon | BB7.2 |
| Konzentration | 5 μL/Test |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16381534
|
eBioscience
63-9876-41 |
25 Tests | |||||||
16391534
![]() |
Invitrogen™
63-9876-42 |
100 Tests |
391.00 €
100 Stück |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with
HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spezifikation
| HLA-A2 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P04439 | |
| HLA-A | |
| Primary | |
| Antibody |
| BB7.2 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Mouse | |
| Human | |
| 3105 | |
| IgG2b κ | |
| 4° C, store in dark, DO NOT FREEZE! | |
| HLA-A |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts