missing translation for 'onlineSavingsMsg'
Learn More

HLA-BC Monoclonal Antibody (B1.23.2), Functional Grade, eBioscience™, Invitrogen™

Artikelnummer. 17151372
Change view
Click to view available options
Menge:
5 mg
25 mg
50 mg
Packungsgröße:
25 Milligramm
5 Milligramm
50 Milligramm
Artikelnummer. Menge unitSize
17151372 25 mg 25 Milligramm
17171372 5 mg 5 Milligramm
17161372 50 mg 50 Milligramm
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17151372

Marke: Affymetrix eBioscience 165935025

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

Description: This B1.23.2 monoclonal antibody recognizes the human Human Leukocyte Antigens (HLA) B and C. Along with HLA-A, these proteins correspond to MHC Class I in humans. HLA-B and HLA-C are expressed on nearly all cell types and heterodimerize with beta-2 microglobulin at the cell surface. These molecular complexes play a critical role in the presentation of endoplasmic reticulum-derived peptides to immune cells. The antibody has been shown to inhibit binding of KIR2DL1 and LIR1. Applications Reported: This B1.23.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B1.23.2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

HLA-BC Monoclonal antibody specifically detects HLA-BC in Human samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen HLA-BC
Anwendungen Flow Cytometry, Functional Assay
Klassifikation Monoclonal
Klon B1.23.2
Konzentration 1 mg/mL
Konjugat Functional Grade
Zusammensetzung PBS with no preservative; pH 7.2
Gen HLA-B
Gen-Zugriffsnummer P01889
Gen-Alias AS; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; epididymis secretory protein Li 83; HEL-S-83; histocompatibility antigen; HLA -B; HLA B-40011; HLA B44; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAB; HLA-B; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B*45ZJ; HLA-B15; HLA-B35; HLA-B35 antigen; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Bw50 antigen; HLA-Cw; HLA-DRB1; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility complex, class I, B; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27d; MHC HLA-B27-HS; MHC HLA-B39N; SPDA1; truncated class I antigen; truncated MHC class I antigen
Wirtsspezies Mouse
Reinigungsverfahren Affinity chromatography
Menge 25 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3106
Zielspezies Human
Inhalt und Lagerung 4° C
Produkttyp Antibody
Form Liquid
Isotype IgG2b κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.