missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA G Antibody (mem-G/9), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-43580
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Spezifikation
| HLA G | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G | |
| The original antibody was generated by immunizing mice with native, properly folded, beta-2-microglubulin (b2m)-associated HLA G. (Uniprot# P17693) | |
| 0.1 mg | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
| 3135 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| mem-G/9 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur