missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP AB Recombinant Rabbit Monoclonal Antibody (PSH05-37)
Beschreibung
hnRNP AB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are produced by RNA polymerase II and are components of the heterogeneous nuclear RNA (hnRNA) complexes. They are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene, which binds to one of the components of the multiprotein editosome complex, has two repeats of quasi-RRM (RNA recognition motif) domains that bind to RNAs. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Spezifikation
Spezifikation
| Antigen | hnRNP AB |
| Anwendungen | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | PSH05-37 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gen | HNRNPAB |
| Gen-Zugriffsnummer | Q99020, Q99729 |
| Gen-Alias | 3010025C11Rik; A1F-C1; ABBP1; ABBP-1; apobec-1 binding protein 1; APOBEC1-binding protein 1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1-binding protein 1; CArG-binding factor-A; Cbf-a; Cgbfa; Heterogeneous nuclear ribonucleoprotein A/B; hnRNP A/B; hnRNP type A/B protein; HNRNPAB; Hnrpab; S1 protein D2 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?