missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (CL11669) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44184
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HO-1/HMOX1/HSP32 Monoclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| HO-1/HMOX1/HSP32 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:200 -1:500 | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| This antibody was generated using a recombinant protein sequence of P09601, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
| 3162 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL11669 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur