missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10701-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.Spezifikation
| HO-1/HMOX1 | |
| Polyclonal | |
| Unconjugated | |
| HMOX1 | |
| bK286B10, heme oxygenase (decycling) 1, Heme oxygenase 1, HMOX1, HO, HO 1, HO1, HSP32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15368, 24451, 3162 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P06762, P09601, P14901 | |
| HMOX1 | |
| HO-1/HMOX1 Fusion Protein Ag1190 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur