Learn More
Homatropine Methylbromide, TRC
Artikelnummer. 30405683
Click to view available options
Menge:
10 g
Packungsgröße:
10 Gramm
Artikelnummer. 30405683
Marke: Toronto Research Chemicals TRCH45922510G
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemischer Name oder Material Homatropine methylbromide
CAS 80-49-9
Summenformel C17 H24 N O3 . Br
Menge 10 g
Synonym Homatropine Methylbromide, (1R,3r,5S)-3-[[(2RS)-2-Hydroxy-2-phenylacetyl]oxy]-8,8-dimethyl-8-azoniabicyclo[3.2.1]octane bromide
InChI-Formel InChI=1S/C17H24NO3.BrH/c1-18(2)13-8-9-14(18)11-15(10-13)21-17(20)16(19)12-6-4-3-5-7-12;/h3-7,13-16,19H,8-11H2,1-2H3;1H/q+1;/p-1/t13-,14+,15+,16-;/m1./s1
SMILES [Br-].C[N+]1(C)[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3
IUPAC-Name [(1S,5R)-8,8-dimethyl-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;bromide
Molekulargewicht (g/mol) 370.28
Formelmasse 369.094