missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB7 Monoclonal Antibody (864617)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561898
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The homeobox genes encode transcription factors that regulate developmental processes such as body patterning and organogenesis. The mammalian HOX gene complex consists of 39 genes that are located on 4 linkage groups (clusters), dispersed over 4 chromosomes (HOXA, HOXB, HOXC and HOXD). HOXB7 is expressed in most malignant T-cell lines and CD4-positive peripheral blood cells, and is constitutively expressed in both melanoma primary lesions and cell lines. HOXB7 binds to and transactivates bFGF in melanoma cell lines but SkBr3 breast adenocarcinoma is negative for the expression of both HOXB7 and bFGF genes. HOXB7 may be a key activator of tumor-associated neoangiogenesis.
Spezifikation
| HOXB7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P09629 | |
| Hoxb7 | |
| E. coli-derived recombinant human HOXB7, Ser2-Asn123. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 864617 | |
| Unconjugated | |
| Hoxb7 | |
| AI325018; HHO.C1; homeo box 2C; homeo box B7; homeo box c1 protein; homeobox B7; homeobox gene B7; homeobox protein HHO.C1; Homeobox protein Hox-2.3; homeobox protein Hox-2C; Homeobox protein Hox-B7; homeobox protein MH-22B; Homeobox protein MuB1; homeobox protein R1B; HOX2; Hox-2.3; Hox-23; HOX2C; Hox2r1b; Hoxb7; Hoxb-7; R1b | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3217 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur