missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp70 interacting protein HIP Antibody (CL3708) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-61615C
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Hsp70 interacting protein HIP Monoclonal antibody specifically detects Hsp70 interacting protein HIP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| Hsp70 interacting protein HIP | |
| Monoclonal | |
| DyLight 650 | |
| AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein | |
| This antibody was developed using a recombinant protein derived from P50502, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Lipid and Metabolism, Membrane Trafficking and Chaperones | |
| 6767 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3708 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur