missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTN1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5122312
This item is not returnable.
View return policy
Description
This gene encodes a member of the histatin family of small, histidine-rich, cationic proteins. They function as antimicrobial peptides and are important components of the innate immune system. Histatins are found in saliva and exhibit antibacterial, antifungal activities and function in wound healing.
Specifications
| HTN1 | |
| Polyclonal | |
| Unconjugated | |
| HTN1 | |
| HIS1; His1 12/38; His1 31/57; His1-(12-38)-peptide; His1-(31-57)-peptide; histatin 1; histatin-1; Histatin-2; Histidine-rich protein 1; HTN1; Post-PB protein; PPB | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3346 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P15515 | |
| HTN1 | |
| Asp20-Asn57 (Accession P15515), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction