missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
This gene encodes a member of the histatin family of small, histidine-rich, cationic proteins. They function as antimicrobial peptides and are important components of the innate immune system. Histatins are found in saliva and exhibit antibacterial, antifungal activities and function in wound healing.
Spezifikation
Spezifikation
| Antigen | HTN1 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gen | HTN1 |
| Gen-Zugriffsnummer | P15515 |
| Gen-Alias | HIS1; His1 12/38; His1 31/57; His1-(12-38)-peptide; His1-(31-57)-peptide; histatin 1; histatin-1; Histatin-2; Histidine-rich protein 1; HTN1; Post-PB protein; PPB |
| Gensymbole | HTN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?