missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS5 Antibody, R&D Systems™

Goat Polyclonal Antibody
201.00 € - 491.00 €
Spezifikation
| Antigen | ADAMTS5 |
|---|---|
| Konzentration | LYOPH |
| Verdünnung | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Anwendungen | Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18755547
![]() |
R&D Systems
AF2198-SP |
25 μg |
201.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18099721
![]() |
R&D Systems
AF2198 |
100 μg |
491.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
ADAMTS5 Polyclonal specifically detects ADAMTS5 in Human samples. It is validated for Western Blot, Immunoprecipitation.Spezifikation
| ADAMTS5 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| A disintegrin and metalloproteinase with thrombospondin motifs 11, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 5 (aggrecanase-2), ADAM metallopeptidase with thrombospondin type 1 motif, 5, ADAM-TS 11, ADAM-TS 5, ADAMTS11, ADAMTS-11, ADAM-TS5, ADAMTS-5, ADMP2, ADMP-2, ADMP-2A disintegrin and metalloproteinase with thrombospondin motifs 5, Aggrecanase 2, Aggrecanase-2, disintegrin-like and metalloprotease with thrombospondin type 1 motif, 510ADAMTS11FLJ36738, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82 | |
| ADAMTS5 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q9UNA0 | |
| 11096 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAMTS5 Ser262-Pro622 Accession # Q9UNA0 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ADAMTS5 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) ADAMTS1 and rhADAMTS-L1.2 is observed. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts