missing translation for 'onlineSavingsMsg'
Learn More

Human AIF-1/Iba1 Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18183031
Change view
Click to view available options
Menge:
100 μg
Unit Size:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18183031 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18183031 Supplier R&D Systems Supplier No. MAB7308

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIF-1/Iba1
Anwendungen Immunohistochemistry
Klassifikation Monoclonal
Klon 603102
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Immunohistochemistry 0.5-25 ug/mL
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer P55008
Gen-Alias AIF1, AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Gensymbole AIF1
Wirtsspezies Mouse
Immunogen E. coli-derived recombinant human AIF-1/Iba1 Met1-Pro147 Accession # P55008
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 199
Testspezifität Detects human AIF-1/Iba1 in direct ELISAs.
Rekonstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.