Learn More
R&D Systems™ Human Amyloid beta (aa1-40) Quantikine ELISA Kit
Beschreibung
The Human Amyloid beta (aa1-40) Quantikine ELISA Kit quantifies human beta Amyloid in cell culture supernates (200 ul), tissue lysates (10 ul), cerebrospinal fluid (10 ul).
Spezifikation
Spezifikation
| Probentyp | Cerebrospinal Fluid, Tissue Lysates |
| Gensymbol | SPEZIAL |
| Synonym | Abeta, Amyloid beta, beta-amyloid peptide |