Learn More
Human B7-1/CD80 Antibody, R&D Systems™
Beschreibung
B7-1/CD80 Polyclonal specifically detects B7-1/CD80 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spezifikation
Spezifikation
| Antigen | B7-1/CD80 |
| Anwendungen | Western Blot, Immunohistochemistry |
| Klassifikation | Polyclonal |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gen-Alias | Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 |
| Gensymbole | CD80 |
| Wirtsspezies | Goat |
| Mehr anzeigen |