missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-2/BMP-4 Antibody, R&D Systems™
Description
BMP-2/BMP-4 Polyclonal specifically detects BMP-2/BMP-4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | BMP-2/BMP-4 |
| Anwendungen | Western Blot, Immunohistochemistry |
| Klassifikation | Polyclonal |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gen-Zugriffsnummer | P12643 |
| Gen-Alias | BDA2, BMP2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, BMP2B, BMP2B1, BMP4, bone morphogenetic protein 2, MCOPS6, OFC11, SSFSC |
| Gensymbole | BMP2 |
| Wirtsspezies | Goat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?