missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BMP-8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marke: R&D Systems AF1073
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BMP-8 Polyclonal specifically detects BMP-8 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spezifikation
| BMP-8 | |
| Polyclonal | |
| Unconjugated | |
| AAP74559 | |
| BMP8A | |
| E. coli-derived recombinant human BMP-8a Ala264-His402 Accession # AAP74559 | |
| 100 μg | |
| Primary | |
| Detects human BMP-8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) BMP-2, rhBMP-3, rhBMP-4, rhBMP-5, rhBMP-6 and rhBMP-7 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| BMP8, BMP8A, BMP-8A, OP-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 353500 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur