missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human C9orf50 Control Fragment Recombinant Protein
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55399 (PA5-55399. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Chromosome 9 consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.
Spezifikation
Spezifikation
| Zugriffsnummer | Q5SZB4 |
| Konzentration | ≥5.0 mg/mL |
| Zur Verwendung mit (Anwendung) | Blocking Assay, Control |
| Zusammensetzung | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 375759 |
| Name | Human C9orf50 Control Fragment |
| Menge | 100 μl |
| Kennzeichnung | RUO |
| Gen-Alias | C9orf50; chromosome 9 open reading frame 50; uncharacterized protein C9orf50 |
| Gebräuchliche Bezeichnung | C9orf50 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur