missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin D Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB10141-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for Immunocytochemistry.
Spezifikation
| Cathepsin D | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin D Leu21-Leu412 Accession # P07339 | |
| 100 μg | |
| 1509 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunocytochemistry | |
| 984824 | |
| Immunocytochemistry 8-25 ug/mL | |
| P07339 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Cathepsin D in direct ELISAs. | |
| Human | |
| Supernatant |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur