missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD58/LFA-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marke: R&D Systems AF1689
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD58/LFA-3 Polyclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Spezifikation
| CD58/LFA-3 | |
| Polyclonal | |
| Unconjugated | |
| P19256 | |
| CD58 | |
| Mouse myeloma cell line NS0-derived recombinant human CD58/LFA-3 Phe29-Arg215 Accession # P19256 | |
| 100 μg | |
| Primary | |
| Detects human CD58/LFA-3 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 3-12 ug/mL | |
| Ag3, CD58, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA3, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 965 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur