missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cdk6 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB11636-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cdk6 Monoclonal antibody specifically detects Cdk6 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,Immunocytochemistry
Spezifikation
| Cdk6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| CDKN6, Cell division protein kinase 6, cyclin-dependent kinase 6, EC 2.7.11, EC 2.7.11.22, PLSTIREMGC59692, Serine/threonine-protein kinase PLSTIRE | |
| E. coli -derived recombinant human CDK-6, Met1-Ala326, Accession # Q00534 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry | |
| 1092920 | |
| Western Blot 0.5 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Q00534 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1021 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur