Learn More
Human CFL1 Antibody, Novus Biologicals Biologicals™
Beschreibung
Cofilin Monoclonal antibody specifically detects Cofilin in Human samples. It is validated for Simple Western,Western Blot,Immunohistochemistry,Immunocytochemistry
Spezifikation
Spezifikation
| Antigen | Cofilin |
| Anwendungen | Simple Western, Western Blot, Immunohistochemistry, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 1093310 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 0.5 μg/mL, Simple Western 10-100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Gen-Zugriffsnummer | P23528 |
| Gen-Alias | 18 kDa phosphoprotein, CFL, cofilin, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, HEL-S-15, p18 |
| Wirtsspezies | Mouse |
| Mehr anzeigen |