missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytokeratin 8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB3165-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cytokeratin 8 Monoclonal specifically detects Cytokeratin 8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Spezifikation
| Cytokeratin 8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| CARD2, CK8, CK-8, CYK8, Cytokeratin-8, EndoA, K2C8, K8, keratin 8, keratin, type II cytoskeletal 8, keratin-8, KO, KRT8, Type-II keratin Kb8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3856 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LP3K | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KRT8 | |
| SV-40 transformed human neonatal keratinocyte extract | |
| 25 μg | |
| Primary | |
| Detects human Cytokeratin 8. | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur