Learn More
Human Desmin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB11607-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Immunocytochemistry,Western Blot,Simple Western,Immunohistochemistry,Multiplex Immunofluorescence
Spezifikation
| Desmin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| CMD1I, CMD1IFLJ41013, CSM1, CSM2, DES, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| E. coli derived recombinant human Desmin, Accession # P17661 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunocytochemistry, Western Blot, Simple Western, Immunohistochemistry, Multiplex | |
| 1089115 | |
| Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL | |
| P17661 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1674 | |
| Human | |
| Purified |