missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DIAPH-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB6806-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DIAPH-2 Monoclonal specifically detects DIAPH-2 in Human samples. It is validated for Western Blot.
Spezifikation
| DIAPH-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| O60879 | |
| DIAPH2 | |
| E. coli-derived recombinant human DIAPH-2 Val314-Gln540 Accession # O60879 | |
| 25 μg | |
| Primary | |
| Detects human DIAPH-2 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 699142 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DIA2, DIAdiaphanous (Drosophila, homolog) 2, DIAPH2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2, POF2FLJ11167, protein diaphanous homolog 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1730 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur