missing translation for 'onlineSavingsMsg'
Learn More

Human FANCG knockout HEK-293T cell lysate, 1 Kit

Artikelnummer. 32155946 Alle ansehen Abcam Produkte
Dieser Artikel kann nicht zurückgegeben werden. View return policy
Product Code. 32155946 Supplier Abcam Supplier No. ab2588701Kit
 Eingeschränkter Artikel
Verfügbar ab: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human FANCG knockout HEK-293T cell/tissue lysate.

  • Species: Human
  • Tissue: FANCG knockout HEK-293T
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.