missing translation for 'onlineSavingsMsg'
Learn More

Human Fc gamma RIIIA/CD16a Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18771417
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18771417 25 μg 25 Mikrogramm
18702707 100 μg 100 Mikrogramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18771417

Marke: R&D Systems MAB107511SP

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

Fc gamma RIIIA/CD16a Monoclonal antibody specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for nan
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Fc gamma RIIIA/CD16a
Klassifikation Monoclonal
Klon 1041646
Konjugat Unconjugated
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gen-Zugriffsnummer P08637
Gen-Alias CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGR3A, FCGRIII, FcgRIIIA, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Wirtsspezies Mouse
Immunogen Mouse myeloma cell line, NS0-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 25 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 2214
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.