Learn More
Human Fibroblast Activation Protein alpha/FAP Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18790669
Change view
Click to view available options
Menge:
100 μg
25 μg
500 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
18790669 100 μg 100 Mikrogramm
18703308 25 μg 25 Mikrogramm
18752519 500 μg 500 Mikrogramm
3 options
Artikelnummer. 18790669
Marke: R&D Systems MAB3715100
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody has been used in 8 publications
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Fibroblast Activation Protein alpha/FAP
Anwendungen Flow Cytometry, CyTOF, ELISA
Klassifikation Monoclonal
Klon 427819
Konjugat Unconjugated
Verdünnung Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer Q12884
Gen-Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
Gensymbole FAP
Wirtsspezies Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 2191
Testspezifität Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed.
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1