Learn More
Human GASP-2/WFIKKN Biotinylated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  R&D Systems BAM21362
Artikelnummer. 18169550
  • 528.00 € / 250 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
GASP-2/WFIKKN Monoclonal specifically detects GASP-2/WFIKKN in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
GASP-2/WFIKKN
Monoclonal
LYOPH
ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Q96NZ8
WFIKKN1
Mouse myeloma cell line NS0-derived recombinant human GASP-2/WFIKKN Ala20-Asp548 Accession # Q96NZ8
250 μg
Primary
Detects human GASP-2/WFIKKN in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human GASP-1 is observed.
Human
Purified
ELISA
289839
Biotin
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
C16orf12, Chromosome 16 Open Reading Frame 12, GASP2, GASP-2, Growth And Differentiation Factor-Associated Serum Protein 2, hGASP-2, RJD2, WAP Four-Disulfide Core Domain 20A, WAP, Follistatin/Kazal, Immunoglobulin, Kunitz And Netrin Domain Containing 1, WAP, FS, Ig, KU, And NTR-Containing Protein, WFDC20A, WFIKKN, WFIKKN1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
117166
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen