missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDF-11/GDF-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB19582
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GDF-11/GDF-8 Monoclonal specifically detects GDF-11/GDF-8 in Human samples. It is validated for ELISA, Neutralization.
Spezifikation
| GDF-11/GDF-8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDF11 | |
| E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390 | |
| 100 μg | |
| Primary | |
| Detects human GDF-11/BMP-11 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with mouse GDF-8 is observed. | |
| Human | |
| Purified |
| Neutralization | |
| 743835 | |
| Neutralization 0.04-0.2 ug/mL | |
| O95390 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10220 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur