Learn More
Human Ghrelin/Obestatin Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marke:  R&D Systems AF8149
Artikelnummer. 18028241
  • 523.00 € / 100 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Ghrelin/Obestatin Polyclonal specifically detects Ghrelin/Obestatin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Bestellinformation
The product is shipped at ambient temperature.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Ghrelin/Obestatin
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, GHRL, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46
Sheep
Antigen Affinity-purified
RUO
51738
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL
Q9UBU3
GHRL
E. coli-derived recombinant human Ghrelin/Obestatin Gly24-Lys117 (Leu72Met) Accession # Q9UBU3
100 μg
Primary
Detects human Ghrelin/Obestatin in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross-reactivity with recombinant mouse Ghrelin/Obestatin is observed.
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen