Learn More
Human GITR Ligand/TNFSF18 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Beschreibung
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry.
Spezifikation
Spezifikation
| Antigen | GITR Ligand/TNFSF18 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 109101 |
| Konzentration | 0.2 mg/mL |
| Konjugat | Alexa Fluor 488 |
| Verdünnung | Flow Cytometry 0.25-1 ug/10^6 cells |
| Zusammensetzung | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gen-Zugriffsnummer | Q9UNG2 |
| Gen-Alias | Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 |
| Mehr anzeigen |
For Research Use Only (RUO)