missing translation for 'onlineSavingsMsg'
Learn More

Human Glucuronosyltransferase 1A1/UGT1A1 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18038621
Change view
Click to view available options
Menge:
100 μg
Tamaño de la unidad:
100 Mikrogramm
Código de producto. Menge unitSize
18038621 100 μg 100 Mikrogramm
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18038621

Marca: R&D Systems AF6490

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

Glucuronosyltransferase 1A1/UGT1A1 Polyclonal specifically detects Glucuronosyltransferase 1A1/UGT1A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen Glucuronosyltransferase 1A1/UGT1A1
Anwendungen Western Blot, Immunohistochemistry, Immunocytochemistry
Klassifikation Polyclonal
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gen-Zugriffsnummer P22309
Gen-Alias bilirubin UDP-glucuronosyltransferase 1-1, bilirubin UDP-glucuronosyltransferase isozyme 1, Bilirubin-specific UDPGT isozyme 1, GNT1, GNT1EC 2.4.1.17, HUG-BR1, UDP glucuronosyltransferase 1 family, polypeptide A1, UDP glycosyltransferase 1 family, polypeptide A1, UDP-glucuronosyltransferase 1-1, UDP-glucuronosyltransferase 1-A, UDP-glucuronosyltransferase 1A1, UDPGT, UDPGT 1-1, UG-BR1, UGT1, UGT1*1, UGT1.1, UGT1-01, UGT1A, UGT-1A, UGT1AUDPGT 1-1, UGT1BILIQTL1
Gensymbole UGT1A1
Wirtsspezies Goat
Immunogen E. coli-derived recombinant human Glucuronosyltransferase 1A1/UGT1A1 Leu60-Thr168 Accession # P22309
Reinigungsverfahren Affinity Purified
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 54658
Testspezifität Detects human Glucuronosyltransferase 1A1/UGT1A1 in direct ELISAs and Western blots.
Rekonstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Zielspezies Human
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.