Learn More
Human Growth Hormone R/GHR Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 2 publications
Marke:  R&D Systems AF1210-SP
Artikelnummer. 18755267
  • ONLINE EXCLUSIVE PRICE
    208.00 €
    196.35 € / 25 Mikrogramm
    Sparen 11.65 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Growth Hormone R Polyclonal specifically detects Growth Hormone R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Growth Hormone R
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
GH receptor, GHBP, GHR, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor
Goat
Affinity Purified
RUO
2690
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 0.125-0.5 ug/mL, CyTOF-ready
P10912
GHR
Mouse myeloma cell line NS0-derived recombinant human Growth Hormone R/GHR Ala27-Tyr264 Accession # P10912
25 μg
Primary
Detects human Growth Hormone R/GHR in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant mouse GHR and recombinant rat GHR is observed.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications