Learn More
Novus Biologicals™ Human Heart Ventricle Whole Tissue Lysate (Adult Whole Arrhythmia)
Artikelnummer. 18210983 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18210983 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18210983 Lieferant Novus Biologicals™ Lieferanten-Nr. NB82059316
Please to purchase this item. Need a web account? Register with us today!
The Human Heart Ventricle Whole Tissue Lysate (Adult Whole Arrhythmia) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Spezifikation
Zur Verwendung mit (Anwendung) Western Blot
Tissue Heart
Menge 1 mg
Lagerungspuffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Testspezifität Total Protein - Arrhythmia, infarct: Heart Ventricle, right
Zelluläre Fraktion Whole
Gewebezustand Arrhythmia
Kreuzreaktivität Human
Wirtsspezies Human
Konzentration 5 mg/mL
Produkttyp Human Heart Ventricle Whole Tissue Lysate (Adult Arrhythmia)
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Lebensphase Erwachs.
Protein State Native
Mehr anzeigen Weniger anzeigen
Nur für Forschungszwecke