Learn More
Gibco™ Human I-309 (CCL1) Recombinant Protein, PeproTech®
Beschreibung
Recombinant Human I-309 is an 8.5 kDa protein consisting of 74 amino acid residues that include six cysteine residues, of which four are the highly conserved cysteine residues present in CC chemokines. Unlike some CC chemokines, whose biologically active form in solution is a non-disulfide-linked dimer, the biologically active form of I-309 is monomeric. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Spezifikation
Spezifikation
| Zugriffsnummer | P22362 |
| Zur Verwendung mit (Anwendung) | ELISA, Functional Assay, Western Blot |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 6346 |
| Molekulargewicht | 8.5 kDa |
| Name | Human I-309 (CCL1) |
| Menge | 1 mg |
| Quelle | E. coli |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <1 EU/ μg |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.