missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human I-TAC (CXCL11) Recombinant Protein, PeproTech®
Click to view available options
Menge:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
20 Mikrogramm
250 Mikrogramm
5 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
Recombinant Human I-TAC is an 8.3 kDa protein containing 73 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interferon-inducible T-cell alpha chemoattractant ( I-TAC), belonging to the CXC chemokine family, is expressed in peripheral blood leukocytes, pancreas and liver, thymus, spleen, lung, small intestine, placenta and prostate. Its molecular weight is 8-11 kD. I-TAC is also known as CXCL-11 and IP-9. The cytokine is involved in regulating calcium release and inducing a chemotactic response in activated T-cells and is the dominant ligand for CXC receptor-3. The gene encoding this protein contains 4 exons and at least three polyadenylation signals which might reflect cell-specific regulation of expression. IFN-gamma is a potent inducer of transcription of this gene. It is upregulated in patients with AIDS, dementia and in patients with multiple sclerosis.
Spezifikation
Spezifikation
| Zugriffsnummer | O14625 |
| Zur Verwendung mit (Anwendung) | ELISA, Functional Assay, Western Blot |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 6373 |
| Molekulargewicht | 8.3 kDa |
| Name | Human I-TAC (CXCL11) |
| Menge | 500 μg |
| Quelle | E. coli |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <1 EU/ μg |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur