Learn More
R&D Systems™ Human IFN-alpha 2 Quantikine HS ELISA Kit
Beschreibung
The Quantikine™ HS Human IFN-alpha 2 Immunoassay is a 4.0 hour solid phase ELISA designed to measure human IFN-alpha 2 (IFN-alpha 2a, IFN-alpha 2b, IFN-alpha 2c) in cell culture supernates, serum, and plasma. It contains HEK293-expressed recombinant human IFN-alpha 2a and has been shown to accurately quantitate the recombinant factor.
Results obtained using natural human IFN-alpha 2 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IFN-alpha 2.
Spezifikation
Spezifikation
| Assay-Bereich | 0.78 to 50 pg/mL |
| Assay-Empfindlichkeit: | 0.107 pg/mL |
| Probentyp | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Spezifität | Natural and recombinant human IFN-alpha 2. |
| Zur Verwendung mit (Anwendung) | ELISA |
| Gen-ID (Entrez) | 3440 |
| Menge | 1 Kit |
| Lagerungsbedingungen | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | alpha-2 A interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2 b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 |
| Ziel | Human |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.