Learn More
Human IFN-beta Antibody, R&D Systems™
Beschreibung
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization.
Spezifikation
Spezifikation
| Antigen | IFN-beta |
| Klassifikation | Monoclonal |
| Klon | 76703R |
| Konjugat | Unconjugated |
| Verdünnung | Neutralization 7-21 ug/mL |
| Zusammensetzung | Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative |
| Gen-Zugriffsnummer | P01574 |
| Gen-Alias | Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
| Gensymbole | IFNB1 |
| Wirtsspezies | Mouse |
| Mehr anzeigen |