Learn More
R&D Systems™ Human IFN-gamma Quantikine ELISA Kit, R&D Systems™
Human IFN-gamma Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Spezifikation
| Assay-Bereich | 15.60 - 1000pg/mL |
|---|---|
| Assay-Empfindlichkeit: | 8pg/mL |
| Probentyp | Cell Culture Supernatant, Plasma (EDTA), Serum |
Beschreibung
The Human IFN-gamma Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IFN-gamma in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul).Spezifikation
| 15.60 - 1000pg/mL | |
| 8pg/mL | |
| Cell Culture Supernatant, Plasma (EDTA), Serum |