missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG Fc APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Marke: R&D Systems FAB110A-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
Specifikationer
| IgG | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IGHG1 | |
| Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region | |
| 100 Tests | |
| Primary | |
| Detects chimeric proteins containing a human IgG Fc region in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 97924 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3500 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering